missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methyl-6-(tri-n-butylstannyl)pyridine, 96%

Brand:  Alfa Aesar™ H51567.03

Additional Details : CAS : 259807-95-9 Weight : 0.00100kg ??product.attributeSet.Transport?? : UN number : 2788 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15418127

  • 293.00 € / 1g


Estimated Shipment: 16-07-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

259807-95-9
382.179
USKQYZHWJZDNCM-UHFFFAOYSA-N
11463183
CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)C
C18H33NSn
MFCD07787401
2-methyl-6-tributylstannyl pyridine, 6-methyl-2-tributylstannyl pyridine, 2-tributylstannyl-6-methylpyridine, 6-methyl-2-tributylstannylpyridine, 6-methyl-2-tributylstannyl-pyridine, pyridine, 2-methyl-6-tributylstannyl, acmc-209gns, amtsn015, 2-methyl-6-tributylstannanyl-pyridine, 6-methylpyridin-2-yl tributylstannane
tributyl-(6-methylpyridin-2-yl)stannane

Specifications

2-Methyl-6-(tri-n-butylstannyl)pyridine
259807-95-9
MFCD07787401
2-methyl-6-tributylstannyl pyridine, 6-methyl-2-tributylstannyl pyridine, 2-tributylstannyl-6-methylpyridine, 6-methyl-2-tributylstannylpyridine, 6-methyl-2-tributylstannyl-pyridine, pyridine, 2-methyl-6-tributylstannyl, acmc-209gns, amtsn015, 2-methyl-6-tributylstannanyl-pyridine, 6-methylpyridin-2-yl tributylstannane
CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)C
382.179
382.16
1g
C18H33NSn
UN2788
USKQYZHWJZDNCM-UHFFFAOYSA-N
tributyl-(6-methylpyridin-2-yl)stannane
11463183
96%
Videos
SDS
Documents
Certificates