Learn More
2-Methyl-6-piperazin-1-ylpyrazine, ≥95%, Thermo Scientific™
Catalog Number p-3175671
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
51047-59-7
C9H14N4
178.239
MFCD08272850
ZQAVHWZTBHGKFK-UHFFFAOYSA-N
20290698
2-methyl-6-piperazin-1-ylpyrazine
CC1=CN=CC(=N1)N2CCNCC2
51047-59-7
C9H14N4
178.239
MFCD08272850
ZQAVHWZTBHGKFK-UHFFFAOYSA-N
20290698
2-methyl-6-piperazin-1-ylpyrazine
CC1=CN=CC(=N1)N2CCNCC2
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2-methyl-6-piperazin-1-yl pyrazine, 2-methyl-6-piperazinylpyrazine, pyrazine,2-methyl-6-1-piperazinyl, pyrazine, 2-methyl-6-1-piperazinyl, pyrazine, 2-methyl-6-1-piperazinyl-9ci
Formula Weight 178.24
Percent Purity 95%
Chemical Name or Material 2-Methyl-6-piperazin-1-ylpyrazine