missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(pentafluorothio)benzyl alcohol, 97%, Thermo Scientific™

Catalog Number 15418375
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1240257-12-8
C8H9F5O2S
264.21
MFCD16652515
HLDZEDSNZDEAMU-UHFFFAOYSA-N
2-methoxy-5-pentafluorosulfur benzyl alcohol, 2-methoxy-5-pentafluoro-??-sulfanyl phenyl methanol, 2-methoxy-5-pentafluorothio benzyl alcohol
66523523
[2-methoxy-5-(pentafluoro-$l^{6}-sulfanyl)phenyl]methanol
COC1=C(C=C(C=C1)S(F)(F)(F)(F)F)CO
This item is not returnable. View return policy
1240257-12-8
C8H9F5O2S
264.21
MFCD16652515
HLDZEDSNZDEAMU-UHFFFAOYSA-N
2-methoxy-5-pentafluorosulfur benzyl alcohol, 2-methoxy-5-pentafluoro-??-sulfanyl phenyl methanol, 2-methoxy-5-pentafluorothio benzyl alcohol
66523523
[2-methoxy-5-(pentafluoro-$l^{6}-sulfanyl)phenyl]methanol
COC1=C(C=C(C=C1)S(F)(F)(F)(F)F)CO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 264.21
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-(pentafluorothio)benzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'