missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3593255
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
868-77-9, 150-76-5
C6H10O3
130.14
MFCD00002863
WOBHKFSMXKNTIM-UHFFFAOYSA-N
2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1
13360
CHEBI:34288
2-hydroxyethyl 2-methylprop-2-enoate
CC(=C)C(=O)OCCO
This item is not returnable. View return policy
868-77-9, 150-76-5
C6H10O3
130.14
MFCD00002863
WOBHKFSMXKNTIM-UHFFFAOYSA-N
2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1
13360
CHEBI:34288
2-hydroxyethyl 2-methylprop-2-enoate
CC(=C)C(=O)OCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 2mg KOH/g max.
CAS Min % 96.0
CAS Max % 100.0
Melting Point -12.0°C
Density 1.0730g/mL
Boiling Point 213°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH2CH2OH
Refractive Index 1.4510 to 1.4530
Quantity 250 g
Beilstein 02,III,1291
Specific Gravity 1.073
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Stabilizer 200 to 400ppm MEHQ
Viscosity 5 mPa.s (30°C)
Formula Weight 130.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Hydroxyethyl methacrylate
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Hydroxyethyl methacrylate, 4-Methoxyphenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'