missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-hydroxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15427346
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1534369-41-9
C12H16BFO3
238.07
MFCD16994234
GXNDNRWVVOKEBG-UHFFFAOYSA-N
2-fluoro-6-hydroxybenzeneboronic acid pinacol ester, 3-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-fluoro-6-hydroxyphenylboronic acid pinacol ester
72219403
3-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
CC1(C)OB(OC1(C)C)C1=C(O)C=CC=C1F
This item is not returnable. View return policy
1534369-41-9
C12H16BFO3
238.07
MFCD16994234
GXNDNRWVVOKEBG-UHFFFAOYSA-N
2-fluoro-6-hydroxybenzeneboronic acid pinacol ester, 3-fluoro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-fluoro-6-hydroxyphenylboronic acid pinacol ester
72219403
3-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
CC1(C)OB(OC1(C)C)C1=C(O)C=CC=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 238.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Fluoro-6-hydroxybenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'