missing translation for 'onlineSavingsMsg'
Learn More

2-[Di(methylthio)methylidene]malononitrile, 97%, Thermo Scientific™

Catalog Number p-2697619
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
5147-80-8
C6H6N2S2
170.248
MFCD00052730
FICQFRCPSFCFBY-UHFFFAOYSA-N
2-bis methylthio methylene malononitrile, bis methylthio methylene malononitrile, 2-di methylthio methylidene malononitrile, propanedinitrile, bis methylthio methylene, 2-bis methylsulfanyl methylidene propanedinitrile, bis methylthio methylene propanedinitrile, 3,3-bis methylthio-2-cyanoacrylonitrile, bis methylthio methylidene propanedinitrile
99229
2-[bis(methylsulfanyl)methylidene]propanedinitrile
CSC(=C(C#N)C#N)SC
This item is not returnable. View return policy
5147-80-8
C6H6N2S2
170.248
MFCD00052730
FICQFRCPSFCFBY-UHFFFAOYSA-N
2-bis methylthio methylene malononitrile, bis methylthio methylene malononitrile, 2-di methylthio methylidene malononitrile, propanedinitrile, bis methylthio methylene, 2-bis methylsulfanyl methylidene propanedinitrile, bis methylthio methylene propanedinitrile, 3,3-bis methylthio-2-cyanoacrylonitrile, bis methylthio methylidene propanedinitrile
99229
2-[bis(methylsulfanyl)methylidene]propanedinitrile
CSC(=C(C#N)C#N)SC
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CN)2C=C(SCH3)2
Quantity 1 g
Formula Weight 170.26
Percent Purity 97%
Chemical Name or Material 2-[Di(methylthio)methylidene]malononitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'