missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite, 95%, Thermo Scientific Chemicals

Catalog Number p-4812794
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
102691-36-1
C15H32N3OP
301.41
RKVHNYJPIXOHRW-UHFFFAOYSA-N
3-bis diisopropylamino phosphino oxy propanenitrile, bis diisopropylamino 2-cyanoethoxy phosphine, 2-cyanoethyl n,n,n',n'-tetraisopropylphosphorodiamidite, 2-cyanoethyl-n,n,n',n'-tetraisopropylphosphorodiamidite, 2-cyanoethyl n,n,n',n'-tetraisopropylphosphordiamidite, phosphorodiamidous acid, tetrakis 1-methylethyl-, 2-cyanoethyl ester, 2-cyanoethyl n,n,n',n'-tetraisopropyl-phosphordiamidite, 2-cyanoethyl tetraisopropylphosphorodiamidite, cetipd, 3-bis diisopropylamino phosphinooxy propanenitrile
128153
3-bis[di(propan-2-yl)amino]phosphanyloxypropanenitrile
CC(C)N(C(C)C)P(N(C(C)C)C(C)C)OCCC#N
This item is not returnable. View return policy
102691-36-1
C15H32N3OP
301.41
RKVHNYJPIXOHRW-UHFFFAOYSA-N
3-bis diisopropylamino phosphino oxy propanenitrile, bis diisopropylamino 2-cyanoethoxy phosphine, 2-cyanoethyl n,n,n',n'-tetraisopropylphosphorodiamidite, 2-cyanoethyl-n,n,n',n'-tetraisopropylphosphorodiamidite, 2-cyanoethyl n,n,n',n'-tetraisopropylphosphordiamidite, phosphorodiamidous acid, tetrakis 1-methylethyl-, 2-cyanoethyl ester, 2-cyanoethyl n,n,n',n'-tetraisopropyl-phosphordiamidite, 2-cyanoethyl tetraisopropylphosphorodiamidite, cetipd, 3-bis diisopropylamino phosphinooxy propanenitrile
128153
3-bis[di(propan-2-yl)amino]phosphanyloxypropanenitrile
CC(C)N(C(C)C)P(N(C(C)C)C(C)C)OCCC#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9490g/mL
Boiling Point 98.0°C to 100.0°C (5.0 mmHg)
Flash Point 60°C
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.47
Quantity 1 g
Specific Gravity 0.949
Formula Weight 301.41
Percent Purity 95%
Chemical Name or Material 2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'