missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloropyrimidine-5-boronic acid pinacol ester, 98%

Brand:  Alfa Aesar™ H64052.MD

Additional Details : CAS : 1003845-08-6 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15418676

  • 55.55 € / 250mg


Estimated Shipment: 23-07-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

1003845-08-6
240.49
VLAPDEKXZLRRKV-UHFFFAOYSA-N
51341943
CC1(C)OB(OC1(C)C)C1=CN=C(Cl)N=C1
C10H14BClN2O2
MFCD11856048
2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidin-5-ylboronic acid pinacol ester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid, pinacol ester, 2-chloropyrimidin-5-yl boronic acid pinacol ester, 2-5-chloro-2-pyrimidyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-chloranyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidine, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

Specifications

2-Chloropyrimidine-5-boronic acid pinacol ester
1003845-08-6
MFCD11856048
2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidin-5-ylboronic acid pinacol ester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid, pinacol ester, 2-chloropyrimidin-5-yl boronic acid pinacol ester, 2-5-chloro-2-pyrimidyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-chloranyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidine, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
CC1(C)OB(OC1(C)C)C1=CN=C(Cl)N=C1
240.49
240.49
98%
250mg
C10H14BClN2O2
15622160
VLAPDEKXZLRRKV-UHFFFAOYSA-N
2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
51341943
Solid
Videos
SDS
Documents
Certificates