missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-(trichloromethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7057080
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1929-82-4
C6H3Cl4N
230.90
MFCD00072500
DCUJJWWUNKIJPH-UHFFFAOYSA-N
2-chloro-6-trichloromethyl pyridine, nitrapyrin, n-serve, pyridine, 2-chloro-6-trichloromethyl, dowco-163, n-serve nitrogen stabilizer, caswell no. 217, nitrapyrine iso-french, unii-8pce86u01w, nitrapyrin ansi:bsi:iso
16004
CHEBI:81935
2-chloro-6-(trichloromethyl)pyridine
ClC1=CC=CC(=N1)C(Cl)(Cl)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1929-82-4
C6H3Cl4N
230.90
MFCD00072500
DCUJJWWUNKIJPH-UHFFFAOYSA-N
2-chloro-6-trichloromethyl pyridine, nitrapyrin, n-serve, pyridine, 2-chloro-6-trichloromethyl, dowco-163, n-serve nitrogen stabilizer, caswell no. 217, nitrapyrine iso-french, unii-8pce86u01w, nitrapyrin ansi:bsi:iso
16004
CHEBI:81935
2-chloro-6-(trichloromethyl)pyridine
ClC1=CC=CC(=N1)C(Cl)(Cl)Cl

2-Chloro-6-(trichloromethyl)pyridine is

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-6-(trichloromethyl)pyridine is

Solubility
Soluble in 10 mg/ml ethanol.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY
Melting Point 60°C to 65°C
Color White
Flash Point 100°C (212°F)
Quantity 1 g
UN Number UN3077
Beilstein 1618997
Solubility Information Soluble in 10mg/ml ethanol.
Formula Weight 230.9
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chloro-6-(trichloromethyl)pyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'