missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7077435
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2401-24-3
C7H8ClNO
157.6
GBOUQGUQUUPGLO-UHFFFAOYSA-N
6-chloro-m-anisidine, 3-amino-4-chloroanisole, benzenamine, 2-chloro-5-methoxy, 2-chloro-5-methoxy-aniline, 6-chloro-m-anisidine hydrochloride, 2-chloro-5-methoxy-phenylamine, 2-chloro-5-methoxybenzenamine hydrochloride, 6-chloro-meta-anisidine hydrochloride, 2-chloro-5-methoxyanilinehydrochloride, timtec-bb sbb008038
75460
2-chloro-5-methoxyaniline
COC1=CC(=C(C=C1)Cl)N
This item is not returnable. View return policy
2401-24-3
C7H8ClNO
157.6
GBOUQGUQUUPGLO-UHFFFAOYSA-N
6-chloro-m-anisidine, 3-amino-4-chloroanisole, benzenamine, 2-chloro-5-methoxy, 2-chloro-5-methoxy-aniline, 6-chloro-m-anisidine hydrochloride, 2-chloro-5-methoxy-phenylamine, 2-chloro-5-methoxybenzenamine hydrochloride, 6-chloro-meta-anisidine hydrochloride, 2-chloro-5-methoxyanilinehydrochloride, timtec-bb sbb008038
75460
2-chloro-5-methoxyaniline
COC1=CC(=C(C=C1)Cl)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 23.0°C to 27.0°C
Density 1.2610g/mL
Boiling Point 135.0°C to 137.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Refractive Index 1.588
Quantity 5 g
Specific Gravity 1.261
Formula Weight 157.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-methoxyaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-5-methoxyaniline, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'