missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitropyridine, 98%, Thermo Scientific™

Catalog Number p-4812776
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
23056-36-2
C5H3ClN2O2
158.54
MFCD00661454
LIEPVGBDUYKPLC-UHFFFAOYSA-N
pyridine, 2-chloro-4-nitro, 2-chloro-4-nitro-pyridine, 2-chlor-4-nitropyridin, pubchem9220, 2-chloro-4nitropyridine, 2chloro-4-nitropyridine, acmc-1ccv7, 2-chloro-4-nitro pyridine, ksc201q1f, 2-chloro-4-nitropyridine
735152
2-chloro-4-nitropyridine
[O-][N+](=O)C1=CC(Cl)=NC=C1
This item is not returnable. View return policy
23056-36-2
C5H3ClN2O2
158.54
MFCD00661454
LIEPVGBDUYKPLC-UHFFFAOYSA-N
pyridine, 2-chloro-4-nitro, 2-chloro-4-nitro-pyridine, 2-chlor-4-nitropyridin, pubchem9220, 2-chloro-4nitropyridine, 2chloro-4-nitropyridine, acmc-1ccv7, 2-chloro-4-nitro pyridine, ksc201q1f, 2-chloro-4-nitropyridine
735152
2-chloro-4-nitropyridine
[O-][N+](=O)C1=CC(Cl)=NC=C1
Melting Point 52.0°C to 56.0°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 158.54
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-nitropyridine
compliance-icons
Product Identifier
  • 2-Chloro-4-nitropyridine, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'