missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methylpyridine-5-boronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7030051
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1010101-07-1
C12H17BClNO2
253.533
MFCD08063084
FQLRMOOIOJBHPS-UHFFFAOYSA-N
2-chloro-3-methylpyridine-5-boronic acid pinacol ester, 2-chloro-3-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-chloro-5-methylpyridine-3-boronic acid pinacol ester, 2-chloro-3-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-chloro-3-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-chloro-5-methylpyridin-3-yl boronic acid pinacol ester
44755178
2-chloro-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)Cl)C
This item is not returnable. View return policy
1010101-07-1
C12H17BClNO2
253.533
MFCD08063084
FQLRMOOIOJBHPS-UHFFFAOYSA-N
2-chloro-3-methylpyridine-5-boronic acid pinacol ester, 2-chloro-3-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-chloro-5-methylpyridine-3-boronic acid pinacol ester, 2-chloro-3-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-chloro-3-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-chloro-5-methylpyridin-3-yl boronic acid pinacol ester
44755178
2-chloro-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)Cl)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 90°C to 93°C
Quantity 250 mg
Formula Weight 253.54
Percent Purity 95%
Chemical Name or Material 2-Chloro-3-methylpyridine-5-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'