missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorophenyl acetic acid, 96%, Thermo Scientific Chemicals

Catalog Number p-7054098
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
739336-26-6
C8H6BrFO2
233.036
MFCD03094440
BPNWLZNAKZSBHH-UHFFFAOYSA-N
2-bromo-5-fluorophenylacetic acid, 2-2-bromo-5-fluorophenyl acetic acid, 2-bromo-5-fluorophenyl acetic acid, 2-bromo-5-fluorophenylaceticacid, 2-bromo-5-fulorophenylacetic acid, benzeneacetic acid, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenyl-acetic acid, pubchem16327, 2-2-bromo-5-fluoro-phenyl acetic acid, acmc-209osx
2773927
2-(2-bromo-5-fluorophenyl)acetic acid
C1=CC(=C(C=C1F)CC(=O)O)Br
This item is not returnable. View return policy
739336-26-6
C8H6BrFO2
233.036
MFCD03094440
BPNWLZNAKZSBHH-UHFFFAOYSA-N
2-bromo-5-fluorophenylacetic acid, 2-2-bromo-5-fluorophenyl acetic acid, 2-bromo-5-fluorophenyl acetic acid, 2-bromo-5-fluorophenylaceticacid, 2-bromo-5-fulorophenylacetic acid, benzeneacetic acid, 2-bromo-5-fluoro, 2-bromo-5-fluoro-phenyl-acetic acid, pubchem16327, 2-2-bromo-5-fluoro-phenyl acetic acid, acmc-209osx
2773927
2-(2-bromo-5-fluorophenyl)acetic acid
C1=CC(=C(C=C1F)CC(=O)O)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 113°C to 114°C
Quantity 1 g
Formula Weight 233.03
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-fluorophenyl acetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'