missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-4-fluorobenzyl bromide, 96%

Brand:  Alfa Aesar™ H62157.06

Additional Details : CAS : 61150-57-0 Weight : 0.00500kg ??product.attributeSet.Transport?? : UN number : 3261 Chem class : 8 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15418728

  • 159.00 € / 5g


Estimated Shipment: 23-07-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

61150-57-0
267.92
QPLUIZXBWYUFMY-UHFFFAOYSA-N
10706977
FC1=CC(Br)=C(CBr)C=C1
C7H5Br2F
MFCD03840524
2-bromo-4-fluorobenzyl bromide, 2-bromo-4-fluorobenzylbromide, 2-bromo-1-bromomethyl-4-fluorobenzene, benzene, 2-bromo-1-bromomethyl-4-fluoro, 4-fluoro-2-bromobenzylbromide, 1-bromo-2-bromomethyl-5-fluorobenzene, pubchem4883, 2-bromo-1-bromomethyl-4-fluoro-benzene, acmc-1b995
2-bromo-1-(bromomethyl)-4-fluorobenzene

Specifications

2-Bromo-4-fluorobenzyl bromide
61150-57-0
MFCD03840524
2-bromo-4-fluorobenzyl bromide, 2-bromo-4-fluorobenzylbromide, 2-bromo-1-bromomethyl-4-fluorobenzene, benzene, 2-bromo-1-bromomethyl-4-fluoro, 4-fluoro-2-bromobenzylbromide, 1-bromo-2-bromomethyl-5-fluorobenzene, pubchem4883, 2-bromo-1-bromomethyl-4-fluoro-benzene, acmc-1b995
FC1=CC(Br)=C(CBr)C=C1
267.92
267.92
96%
5g
C7H5Br2F
UN3261
QPLUIZXBWYUFMY-UHFFFAOYSA-N
2-bromo-1-(bromomethyl)-4-fluorobenzene
10706977
Solid
Videos
SDS
Documents
Certificates