missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-cyclopropyl-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.
Solubility
Soluble in Methanol. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container. Store at -20°C.
2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.
Solubility
Soluble in Methanol. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container. Store at -20°C.
Specifications
Melting Point | 212°C to 216°C |
Quantity | 1 g |
Beilstein | 124356 |
Solubility Information | Soluble in Methanol. Insoluble in water. |
Formula Weight | 141.2 |
Percent Purity | 98% |
Chemical Name or Material | 2-Amino-5-cyclopropyl-1,3,4-thiadiazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'