missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloropyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-4258745
39.06 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
19798-80-2
C5H6ClN2
129.57
MFCD04113820
RQMWVVBHJMUJNZ-UHFFFAOYSA-O
2-amino-4-chloropyridine, 4-chloro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro, 4-chloro-2-pyridinamine, 4-chloro-2-pyridylamine, 2-amino-4-chloro pyridine, 4-chloro-2-pyridinylamine, 2-amino-4-chloro-pyridine, 4-chloro-2-aminopyridine, 4-chlorpyridin-2-amin
581866
4-chloropyridin-2-amine
NC1=CC(Cl)=CC=[NH+]1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
19798-80-2
C5H6ClN2
129.57
MFCD04113820
RQMWVVBHJMUJNZ-UHFFFAOYSA-O
2-amino-4-chloropyridine, 4-chloro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro, 4-chloro-2-pyridinamine, 4-chloro-2-pyridylamine, 2-amino-4-chloro pyridine, 4-chloro-2-pyridinylamine, 2-amino-4-chloro-pyridine, 4-chloro-2-aminopyridine, 4-chlorpyridin-2-amin
581866
4-chloropyridin-2-amine
NC1=CC(Cl)=CC=[NH+]1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 130.0°C to 132.0°C
Color Brown to Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 128.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Amino-4-chloropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-4-chloropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'