missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2'-chloro-5-nitrobenzophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-7057094
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2011-66-7
C13H9ClN2O3
276.68
MFCD00792453
GRDGBWVSVMLKBV-UHFFFAOYSA-N
2-amino-2'-chloro-5-nitrobenzophenone, 2-amino-5-nitrophenyl 2-chlorophenyl methanone, 2-amino-5-nitro-2'-chlorobenzophenone, 2-amino-2'-chloro-5-nitro benzophenone, 2-2-chlorobenzoyl-4-nitroaniline, methanone, 2-amino-5-nitrophenyl 2-chlorophenyl, 2-amino-5-nitrophenyl-2-chlorophenyl methanone, 2-amino-5-nitro-phenyl-2-chloro-phenyl-methanone, 2-amino-5-nitro-2'-chloro benzophenone, pubchem3255
74830
(2-amino-5-nitrophenyl)-(2-chlorophenyl)methanone
NC1=CC=C(C=C1C(=O)C1=CC=CC=C1Cl)[N+]([O-])=O
This item is not returnable. View return policy
2011-66-7
C13H9ClN2O3
276.68
MFCD00792453
GRDGBWVSVMLKBV-UHFFFAOYSA-N
2-amino-2'-chloro-5-nitrobenzophenone, 2-amino-5-nitrophenyl 2-chlorophenyl methanone, 2-amino-5-nitro-2'-chlorobenzophenone, 2-amino-2'-chloro-5-nitro benzophenone, 2-2-chlorobenzoyl-4-nitroaniline, methanone, 2-amino-5-nitrophenyl 2-chlorophenyl, 2-amino-5-nitrophenyl-2-chlorophenyl methanone, 2-amino-5-nitro-phenyl-2-chloro-phenyl-methanone, 2-amino-5-nitro-2'-chloro benzophenone, pubchem3255
74830
(2-amino-5-nitrophenyl)-(2-chlorophenyl)methanone
NC1=CC=C(C=C1C(=O)C1=CC=CC=C1Cl)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 119°C to 123°C
Color Yellow
Quantity 5 g
Beilstein 2814216
Sensitivity Air sensitive
Formula Weight 276.68
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-2'-chloro-5-nitrobenzophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'