missing translation for 'onlineSavingsMsg'
Learn More

2,6-Lutidine N-oxide, 98%

Product Code. p-7021811
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11452477

Brand: Thermo Scientific Alfa Aesar A14299.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1073-23-0 | C7H9NO | 123.155 g/mol

2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with oxidizing agents, moisture and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073-23-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00039715
InChI Key LIDGFHXPUOJZMK-UHFFFAOYSA-N
Synonym 2,6-dimethylpyridine n-oxide, 2,6-lutidine n-oxide, 2,6-lutidine-n-oxide, 2,6-dimethylpyridine 1-oxide, 2,6-lutidine 1-oxide, pyridine, 2,6-dimethyl-, 1-oxide, 2,6-lutidine oxide, 2,6-dimethylpyridine oxide, 2,6-dimethylpyridinium n-oxide, 2,6-dimethylpyridin-1-ium-1-olate
PubChem CID 14083
IUPAC Name 2,6-dimethyl-1-oxidopyridin-1-ium
SMILES CC1=[N+](C(=CC=C1)C)[O-]

Specifications

Density 1.102
Boiling Point 108°C to 111°C (10 mmHg)
Refractive Index 1.5685
Quantity 10 g
Beilstein 110551
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 2,6-Lutidine N-oxide
compliance-icons
Product Identifier
  • 2,6-Lutidine N-oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,6-Lutidine N-oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.