missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dinitrotoluene, 97%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 58°C to 65°C |
| Boiling Point | 285°C |
| Flash Point | 207°C (404°F) |
| Quantity | 5 g |
| UN Number | UN3454 |
| Beilstein | 2052046 |
| Formula Weight | 182.14 |
| Percent Purity | 97% |
| Chemical Name or Material | 2,6-Dinitrotoluene |
Safety and Handling
Product Identifier
- 2,6-Dinitrotoluene
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : 2,6-Dinitrotoluene
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction