missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzyl chloride, 98%, Thermo Scientific™

Catalog Number 15525976
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
5402-60-8
C9H11Cl
154.64
HPVRFWQMBYLJRL-UHFFFAOYSA-N
2,6-dimethylbenzyl chloride, 2-chloromethyl-1,3-dimethylbenzene, 2,6-dimethylbenzylchloride, 2,6-dimethyl benzylchloride, 1-chloromethyl-2,6-dimethylbenzene, 2-chloromethyl-1,3-dimethyl-benzene, 2-chloromethyl-m-xylene, benzene, 2-chloromethyl-1,3-dimethyl, 2, 6-dimethylbenzyl chloride
221154
2-(chloromethyl)-1,3-dimethylbenzene
CC1=C(C(=CC=C1)C)CCl
This item is not returnable. View return policy
5402-60-8
C9H11Cl
154.64
HPVRFWQMBYLJRL-UHFFFAOYSA-N
2,6-dimethylbenzyl chloride, 2-chloromethyl-1,3-dimethylbenzene, 2,6-dimethylbenzylchloride, 2,6-dimethyl benzylchloride, 1-chloromethyl-2,6-dimethylbenzene, 2-chloromethyl-1,3-dimethyl-benzene, 2-chloromethyl-m-xylene, benzene, 2-chloromethyl-1,3-dimethyl, 2, 6-dimethylbenzyl chloride
221154
2-(chloromethyl)-1,3-dimethylbenzene
CC1=C(C(=CC=C1)C)CCl
Melting Point 33.0°C
Boiling Point 100°C (2.0 kPa)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Formula Weight 154.64
Percent Purity 98%
Chemical Name or Material 2,6-Dimethylbenzyl chloride
compliance-icons
Product Identifier
  • 2,6-Dimethylbenzyl chloride, 98%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'