missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7027675
28.66 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
100379-00-8
C8H11BO2
149.98
MFCD01009693
ZXDTWWZIHJEZOG-UHFFFAOYSA-N
2,6-dimethylphenyl boronic acid, 2,6-dimethylbenzeneboronic acid, 2,6-dimethylphenyl boranediol, 2,6-dimethylboronic acid, m-xylene-2-boronic acid, 2,6-dimethyl-phenylboronic acid, boronic acid, 2,6-dimethylphenyl, 2-borono-m-xylene, pubchem16357, acmc-1c3za
583322
(2,6-dimethylphenyl)boronic acid
CC1=CC=CC(C)=C1B(O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
100379-00-8
C8H11BO2
149.98
MFCD01009693
ZXDTWWZIHJEZOG-UHFFFAOYSA-N
2,6-dimethylphenyl boronic acid, 2,6-dimethylbenzeneboronic acid, 2,6-dimethylphenyl boranediol, 2,6-dimethylboronic acid, m-xylene-2-boronic acid, 2,6-dimethyl-phenylboronic acid, boronic acid, 2,6-dimethylphenyl, 2-borono-m-xylene, pubchem16357, acmc-1c3za
583322
(2,6-dimethylphenyl)boronic acid
CC1=CC=CC(C)=C1B(O)O

It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 120°C to 126°C (decomposition)
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 149.99
Percent Purity 97%
Chemical Name or Material 2,6-Dimethylbenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'