missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(4-fluorobenzoyl)pyrimidine, 95%, Thermo Scientific Chemicals

Catalog Number p-7031335
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1099597-81-5
MFCD11100517
This item is not returnable. View return policy
1099597-81-5
MFCD11100517

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 95°C to 99°C
Molecular Formula C11H5Cl2FN2O
Quantity 250 mg
Formula Weight 271.08
Percent Purity 95%
Chemical Name or Material 2,6-Dichloro-4-(4-fluorobenzoyl)pyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'