Learn More
2,4-Dimethyl-3-furoic acid, 97%, Thermo Scientific™
Catalog Number p-4391324
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
15058-72-7
C7H8O3
140.138
JFXXYKVSLHRBAA-UHFFFAOYSA-N
2,4-dimethyl-3-furoic acid, 2,4-dimethyl-3-furancarboxylic acid, 3-furancarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-furan-3-carboxylic acid, 2,4-dimethylfurane-3-carboxylic acid
638456
2,4-dimethylfuran-3-carboxylic acid
CC1=COC(=C1C(=O)O)C
15058-72-7
C7H8O3
140.138
JFXXYKVSLHRBAA-UHFFFAOYSA-N
2,4-dimethyl-3-furoic acid, 2,4-dimethyl-3-furancarboxylic acid, 3-furancarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-furan-3-carboxylic acid, 2,4-dimethylfurane-3-carboxylic acid
638456
2,4-dimethylfuran-3-carboxylic acid
CC1=COC(=C1C(=O)O)C
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 2, 4-Dimethyl-3-furoic acid