missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)benzimidazole, 95%, Thermo Scientific™

Catalog Number p-7056067
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
2622-74-4
C13H9BrN2
273.13
MFCD00224360
YRWMGOSKROWAIT-UHFFFAOYSA-N
2-4-bromophenyl benzimidazole, 2-4-bromophenyl-1h-benzo d imidazole, 2-4-bromophenyl-1h-benzimidazole, 2-4-bromophenyl-1h-benzoimidazole, 2-4-bromophenyl benzoimidazole, 2-4-bromo-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-4-bromophenyl, 2-4-bromophenyl-1h-1,3-benzodiazole, acmc-209gpy, boron-mol bm885
286182
2-(4-bromophenyl)-1H-benzimidazole
BrC1=CC=C(C=C1)C1=NC2=CC=CC=C2N1
This item is not returnable. View return policy
2622-74-4
C13H9BrN2
273.13
MFCD00224360
YRWMGOSKROWAIT-UHFFFAOYSA-N
2-4-bromophenyl benzimidazole, 2-4-bromophenyl-1h-benzo d imidazole, 2-4-bromophenyl-1h-benzimidazole, 2-4-bromophenyl-1h-benzoimidazole, 2-4-bromophenyl benzoimidazole, 2-4-bromo-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-4-bromophenyl, 2-4-bromophenyl-1h-1,3-benzodiazole, acmc-209gpy, boron-mol bm885
286182
2-(4-bromophenyl)-1H-benzimidazole
BrC1=CC=C(C=C1)C1=NC2=CC=CC=C2N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 162670
Formula Weight 273.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(4-Bromophenyl)benzimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'