missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number p-7054440
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1073339-17-9
C12H15BF2O2
240.06
MFCD12405347
BFCCQFHEIXEEKP-UHFFFAOYSA-N
2-2,3-difluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2,3-difluorophenylboronic acid pinacol ester, 2,3-difluorobenzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-2,3-difluorophenyl-4,4,5,5-tetramethyl
56973143
2-(2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=C(F)C(F)=CC=C1
This item is not returnable. View return policy
1073339-17-9
C12H15BF2O2
240.06
MFCD12405347
BFCCQFHEIXEEKP-UHFFFAOYSA-N
2-2,3-difluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2,3-difluorophenylboronic acid pinacol ester, 2,3-difluorobenzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-2,3-difluorophenyl-4,4,5,5-tetramethyl
56973143
2-(2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=C(F)C(F)=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 240.05
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2,3-Difluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'