missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromosuccinic acid, 98%, Thermo Scientific™

Catalog Number 15426778
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
526-78-3
C4H4Br2O4
275.88
MFCD00004209
FJWGRXKOBIVTFA-UHFFFAOYSA-N
2,3-dibromosuccinic acid, butanedioic acid, 2,3-dibromo, 2,3-dibromosuccinicacid, sym.-dibromosuccinic acid, acmc-20a6bk, succinic acid,3-dibromo, 2,3-dibromo succinic acid, butanedioic acid,3-dibromo, 2,3-dibromosuccinic acid #, 2,3-dibromo-butanedioic acid
95324
2,3-dibromobutanedioic acid
C(C(C(=O)O)Br)(C(=O)O)Br
This item is not returnable. View return policy
526-78-3
C4H4Br2O4
275.88
MFCD00004209
FJWGRXKOBIVTFA-UHFFFAOYSA-N
2,3-dibromosuccinic acid, butanedioic acid, 2,3-dibromo, 2,3-dibromosuccinicacid, sym.-dibromosuccinic acid, acmc-20a6bk, succinic acid,3-dibromo, 2,3-dibromo succinic acid, butanedioic acid,3-dibromo, 2,3-dibromosuccinic acid #, 2,3-dibromo-butanedioic acid
95324
2,3-dibromobutanedioic acid
C(C(C(=O)O)Br)(C(=O)O)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 274°C
Linear Formula HOOCCHBrCHBrCOOH
Quantity 25 g
UN Number UN3261
Formula Weight 275.88
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,3-Dibromosuccinic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'