missing translation for 'onlineSavingsMsg'
Learn More

2-[3,5-di(tert-butyl)-4-hydroxyphenyl]acetic acid, Thermo Scientific™

Catalog Number p-4391239
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
1611-03-6
C16H24O3
264.365
QLMGIWHWWWXXME-UHFFFAOYSA-N
3,5-di-tert-butyl-4-hydroxyphenylacetic acid, 3,5-di-tert-butyl-4-hydroxyphenyl acetic acid, 2-3,5-di tert-butyl-4-hydroxyphenyl acetic acid, 2-3,5-di-tert-butyl-4-hydroxyphenyl acetic acid, benzeneacetic acid,3,5-bis 1,1-dimethylethyl-4-hydroxy, acmc-1bvft, 4-hydroxy-3,5-di-t-butylphenylacetic acid, 3,5-di-t-butyl-4-hydroxyphenyl-acetic acid, 4-hydroxy-3,5-di-t-butyl-phenylacetic acid, 3,5-di-tert-butyl-4-hydroxybenzeneacetic acid
2775110
2-(3,5-ditert-butyl-4-hydroxyphenyl)acetic acid
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC(=O)O
This item is not returnable. View return policy
1611-03-6
C16H24O3
264.365
QLMGIWHWWWXXME-UHFFFAOYSA-N
3,5-di-tert-butyl-4-hydroxyphenylacetic acid, 3,5-di-tert-butyl-4-hydroxyphenyl acetic acid, 2-3,5-di tert-butyl-4-hydroxyphenyl acetic acid, 2-3,5-di-tert-butyl-4-hydroxyphenyl acetic acid, benzeneacetic acid,3,5-bis 1,1-dimethylethyl-4-hydroxy, acmc-1bvft, 4-hydroxy-3,5-di-t-butylphenylacetic acid, 3,5-di-t-butyl-4-hydroxyphenyl-acetic acid, 4-hydroxy-3,5-di-t-butyl-phenylacetic acid, 3,5-di-tert-butyl-4-hydroxybenzeneacetic acid
2775110
2-(3,5-ditert-butyl-4-hydroxyphenyl)acetic acid
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC(=O)O
Quantity 10 g
Formula Weight 264.36
Percent Purity 97%
Chemical Name or Material 2-[3, 5-di(tert-butyl)-4-hydroxyphenyl]acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'