missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoropyridine-4-acetic acid, 98%, Thermo Scientific™

Catalog Number p-7032360
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
65697-73-6
C7H3F4NO2
209.1
MFCD02944320
SOHLFVREHOCIMD-UHFFFAOYSA-N
2,3,5,6-tetrafluoro-4-pyridinyl acetic acid, 2,3,5,6-tetrafluoropyridine-4-yl acetic acid, 2-perfluoropyridin-4-yl acetic acid, 2,3,5,6-tetrafluoropyridine-4-acetic acid, 2,3,5,6-tetrafluoropyridin-4-yl acetic acid, tetrafluoropyridin-4-yl acetic acid, 2-2,3,5,6-tetrafluoropyridin-4-yl acetic acid
1237119
2-(2,3,5,6-tetrafluoropyridin-4-yl)acetic acid
C(C1=C(C(=NC(=C1F)F)F)F)C(=O)O
This item is not returnable. View return policy
65697-73-6
C7H3F4NO2
209.1
MFCD02944320
SOHLFVREHOCIMD-UHFFFAOYSA-N
2,3,5,6-tetrafluoro-4-pyridinyl acetic acid, 2,3,5,6-tetrafluoropyridine-4-yl acetic acid, 2-perfluoropyridin-4-yl acetic acid, 2,3,5,6-tetrafluoropyridine-4-acetic acid, 2,3,5,6-tetrafluoropyridin-4-yl acetic acid, tetrafluoropyridin-4-yl acetic acid, 2-2,3,5,6-tetrafluoropyridin-4-yl acetic acid
1237119
2-(2,3,5,6-tetrafluoropyridin-4-yl)acetic acid
C(C1=C(C(=NC(=C1F)F)F)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 145°C to 149°C
Quantity 250 mg
Formula Weight 209.1
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrafluoropyridine-4-acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'