missing translation for 'onlineSavingsMsg'
Learn More

2,3,3-Trimethylindolenine, 98%, Thermo Scientific Chemicals

Catalog Number p-7026420
21.06 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1640-39-7
C11H13N
159.232
MFCD00005724
FLHJIAFUWHPJRT-UHFFFAOYSA-N
2,3,3-trimethylindolenine, 2,3,3-trimethyl-3h-indole, 3h-indole, 2,3,3-trimethyl, ccris 6607, 2,3,3-trimethyl-indolenin, 2,3,3-trimethyl-3-hydroindole, pubchem7445, rarechem ah bs 0130, 2,3-trimethylindolenine, 2,3,3-trimethyl-indole
15427
2,3,3-trimethylindole
CC1=NC2=CC=CC=C2C1(C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1640-39-7
C11H13N
159.232
MFCD00005724
FLHJIAFUWHPJRT-UHFFFAOYSA-N
2,3,3-trimethylindolenine, 2,3,3-trimethyl-3h-indole, 3h-indole, 2,3,3-trimethyl, ccris 6607, 2,3,3-trimethyl-indolenin, 2,3,3-trimethyl-3-hydroindole, pubchem7445, rarechem ah bs 0130, 2,3-trimethylindolenine, 2,3,3-trimethyl-indole
15427
2,3,3-trimethylindole
CC1=NC2=CC=CC=C2C1(C)C

It is used in synthesis of cyanine dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in synthesis of cyanine dyes.

Solubility
Soluble in chloroform, toluene or dichlorobenzene.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 6°C to 8°C
Density 0.992
Boiling Point 228°C to 229°C
Flash Point 93°C (199°F)
Refractive Index 1.547
Quantity 5 g
Beilstein 119740
Solubility Information Soluble in chloroform,toluene or dichlorobenzene.
Formula Weight 159.23
Percent Purity 98%
Chemical Name or Material 2,3,3-Trimethylindolenine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'