missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Naphthyl)indole, 98%, Thermo Scientific Chemicals

Catalog Number p-7025102
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
23746-81-8
C18H13N
243.31
MFCD00124144
CACDYUNTCMPDMI-UHFFFAOYSA-N
2-2-naphthyl indole, 2-2-naphthyl-1h-indole, 2-naphthalen-2-yl-1h-indole, acmc-1chvp, 2-2-naphthyl-indol, maybridge1_005904, 2-2-indolyl naphthalene, 2-1h-indol-2-yl naphthalene, 2-2-naphthalenyl-1h-indole
265232
N1C2=CC=CC=C2C=C1C1=CC=C2C=CC=CC2=C1
This item is not returnable. View return policy
23746-81-8
C18H13N
243.31
MFCD00124144
CACDYUNTCMPDMI-UHFFFAOYSA-N
2-2-naphthyl indole, 2-2-naphthyl-1h-indole, 2-naphthalen-2-yl-1h-indole, acmc-1chvp, 2-2-naphthyl-indol, maybridge1_005904, 2-2-indolyl naphthalene, 2-1h-indol-2-yl naphthalene, 2-2-naphthalenyl-1h-indole
265232
N1C2=CC=CC=C2C=C1C1=CC=C2C=CC=CC2=C1

It is used in the catalytic enantioselective addition of indoles to arylnitroalkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the catalytic enantioselective addition of indoles to arylnitroalkenes.

Solubility
Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Solubility Information Soluble in DMSO.
IUPAC Name 2-naphthalen-2-yl-1H-indole
Formula Weight 243.31
Percent Purity 98%
Chemical Name or Material 2-(2-Naphthyl)indole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'