Learn More
Avantor™ J.T. Baker™, BAKERBOND spe™ SPE columns Activated Spherical Carbon
Disposable PP or glass columns pre-packed with high capacity sorbents contained between two 20 μm PE frits
Brand:  Avantor™ 7575-06
294.50 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Additional Details : Weight : 0.40000kg
Product Code. 15577684
  • 310.00 € / Each
Estimated Shipment: 18-07-2024
to see stock.
Add to basket
Description
Description
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred μL up to 1 L
  • Processing by vacuum, positive pressure or centrifugation
  • General applications: Extraction and purification of organic compounds such as acrylamide and other polar compounds.
Specifications
Specifications
6 mL
500 mg
30
Activated spherical carbon
40 μm