Learn More
Julabo™ Aqua Stabil™ Water Bath Protective Media
Julabo™ Aqua Stabil™ Water Bath Protective Media stops growth of algae, bacteria and other microorganisms with its bactericidal effectiveness.
Brand: Julabo™ 8940006
Additional Details : Weight : 0.75240kg
Description
- Protects bath fluid
- Provides long period of use for fluid
Specifications
6 Bottles | |
ED, ED-13, ED-13A, ED-13M, ED-17 Open heating bath circulator, Water bath | |
Aqua Stabil Protective Media |
Safety and Handling
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : poly(hexamethylenebiguanide) hydrochloride