missing translation for 'onlineSavingsMsg'
Learn More

250GR Sulfamic Acid BAKER ANALYZED Reagent

Brand:  JT Baker 1828.0250

161.50 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.



250GR Sulfamic Acid BAKER ANALYZED Reagent

Additional Details : CAS : 5329-14-6 Weight : 0.25000kg ??product.attributeSet.Transport?? : UN number : 2967 Chem class : 8 Pack group : III


Product Code. 10645442

  • 170.00 € / 250g
Estimated Shipment: 17-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362-Take off contaminated clothing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Promotions

Promotions