Learn More
1LT HYDRA-POINT Titrant 5mg H2O/ml BAKER ANALYZEDReagent
Brand:  JT Baker 8844.1000
129.20 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
1LT HYDRA-POINT Titrant 5mg H2O/ml BAKER ANALYZEDReagent
Additional Details : Weight : 0.86700kg ??product.attributeSet.Transport?? : UN number : 3286 Chem class : 3 Pack group : II
Product Code. 10599221
  • 136.00 € / 1 L
Estimated Shipment: 05-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.