missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millipore Proteinase K
From Tritirachium album
Brand: Merck Millipore 1073930010
Additional Details : Weight : 0.01000kg
Description
- Solution in Tris/HCl pH 7.5
- 0.01 mol/L
- 600 mAnson-U/mL
- For molecular biology
- EC 3.4.21.14
- Appearance (colour): colourless
- Appearance (clearness): clear
- Activity (hemoglobin; pH 7.5; 37°C): ≥ 600 m Anson U/mL
- Spec. activity (calc. on protein): ≥ 40.0 m Anson U/mg
- DNases (Nicking activity; pBR 322; 6 h; 37 °C): not detectable
- RNases (RNA; 2 h; 37°C): not detectable
- Colony count (aerobic bacteria): ≤ 10 CFU/mL
Specifications
7.5 (25°C in H2O, undiluted) | |
2°C to 8°C | |
10 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Proteinase K
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Proteinase K