missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, BAKER, J.T. Baker™

Catalog Number 10103790
Click to view available options
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Meets Reagent Specifications for testing Ph. Eur. monographs

CAS 77-92-9
Melting Point 149°C to 151°C
Boiling Point 310°C (1013 hPa)
Appearance Passes Test
Assay Percent Range 99.5 to 101.0%
Molecular Formula C6H8O7
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 192.12
Density 1.665 g/cm³ (20°C)
Quantity 1 kg
Packaging Plastic bottle for solids
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'