Learn More
(+/-)-1-Octyn-3-ol, 98%
Product Code. 11474150
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. 11474150
Brand: Thermo Scientific Alfa Aesar A13267.30
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 818-72-4 | C8H14O | 126.20 g/mol
1-Octyn-3-ol was used in the synthesis of synthetic tricolorin A which is an organic chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Octyn-3-ol was used in the synthesis of synthetic tricolorin A which is an organic chemical.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 818-72-4
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00004588
InChI Key VUGRNZHKYVHZSN-UHFFFAOYNA-N
Synonym 1-octyn-3-ol, 1-octyne-3-ol, oct-1-yne-3-ol, 1-oct-1-yn-3-ol, 3-hydroxyoct-1-yne, acmc-20apfv, acmc-20apkf, rac-1-octyn-3-ol, oct-1 yn-3 ol, pubchem11033
PubChem CID 13166
IUPAC Name oct-1-yn-3-ol
SMILES CCCCCC(O)C#C
Specifications
Melting Point -60°C
Density 0.865
Boiling Point 180°C to 181°C
Flash Point 63°C (145°F)
Refractive Index 1.442
Quantity 250 g
Beilstein 1098642
Solubility Information Soluble in water.
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material (+/-)-1-Octyn-3-ol