missing translation for 'onlineSavingsMsg'
Learn More

1-O-n-Octyl-β-D-glucopyranoside, 98%, Thermo Scientific Chemicals

Catalog Number p-3737095
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
29836-26-8
C14H28O6
292.37
MFCD00063288
HEGSGKPQLMEBJL-RKQHYHRCSA-N
octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside
62852
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
This item is not returnable. View return policy
29836-26-8
C14H28O6
292.37
MFCD00063288
HEGSGKPQLMEBJL-RKQHYHRCSA-N
octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside
62852
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 105°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 6857
Specific Rotation Condition − 32.00 (20.00°C c=5,water)
Specific Rotation -32
Solubility Information (10% in water) Clear to slightly hazy colorless to light yellow
Formula Weight 292.36
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1-O-n-Octyl-β-D-glucopyranoside
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'