missing translation for 'onlineSavingsMsg'
Learn More

1-Methylimidazole-4-carboxylic acid, 95%, Thermo Scientific™

Catalog Number 15418357
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
41716-18-1
C5H6N2O2
126.115
MFCD02179560
WZTRQGJMMHMFGH-UHFFFAOYSA-N
1-methyl-1h-imidazole-4-carboxylic acid, 1-methyl-imidazole-4-carboxylic acid, 1-methyl-4-imidazole-carboxylic acid, 1-methyl-4-imidazolecarboxylicacid, pubchem9001, acmc-1am6x, 1-methyl-4-imidazolic acid, ksc237q3j, 1-methyl-4-imidazolecarboxylic acid, 1-methyl-1h-4carboxylic acid
541509
CHEBI:74746
1-methylimidazole-4-carboxylic acid
CN1C=C(N=C1)C(=O)O
This item is not returnable. View return policy
41716-18-1
C5H6N2O2
126.115
MFCD02179560
WZTRQGJMMHMFGH-UHFFFAOYSA-N
1-methyl-1h-imidazole-4-carboxylic acid, 1-methyl-imidazole-4-carboxylic acid, 1-methyl-4-imidazole-carboxylic acid, 1-methyl-4-imidazolecarboxylicacid, pubchem9001, acmc-1am6x, 1-methyl-4-imidazolic acid, ksc237q3j, 1-methyl-4-imidazolecarboxylic acid, 1-methyl-1h-4carboxylic acid
541509
CHEBI:74746
1-methylimidazole-4-carboxylic acid
CN1C=C(N=C1)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 241°C to 243°C
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 126.11
Percent Purity 95%
Chemical Name or Material 1-Methylimidazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'