missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexyne, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
1-Hexyne is used in the preparation of tricyclic isoindolinone scaffold by undergoing hydrozirconation and ring-closing metathesis. It is used to prepare trans-1,2-bis(5-thianthreniumyl)alkene tetrafluoroborate by reacting with thianthrene cation radical tetrafluoroborate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Hexyne is used in the preparation of tricyclic isoindolinone scaffold by undergoing hydrozirconation and ring-closing metathesis. It is used to prepare trans-1,2-bis(5-thianthreniumyl)alkene tetrafluoroborate by reacting with thianthrene cation radical tetrafluoroborate.
Solubility
Miscible with organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
1-Hexyne is used in the preparation of tricyclic isoindolinone scaffold by undergoing hydrozirconation and ring-closing metathesis. It is used to prepare trans-1,2-bis(5-thianthreniumyl)alkene tetrafluoroborate by reacting with thianthrene cation radical tetrafluoroborate.
Solubility
Miscible with organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
Specifications
Melting Point | -132°C |
Density | 0.718 |
Boiling Point | 69°C to 71°C |
Flash Point | −21°C (−5°F) |
Odor | Characteristic |
Refractive Index | 1.399 |
Quantity | 10 g |
UN Number | UN3295 |
Beilstein | 635687 |
Solubility Information | Miscible with organic solvents. Immiscible with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'