missing translation for 'onlineSavingsMsg'
Learn More

1-Fmoc-piperidine-4-thiocarboxamide, 97%, Thermo Scientific™

Catalog Number p-7039317
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
941717-12-0
C21H22N2O2S
366.479
MFCD22988962
URQBPQRQAPSGOQ-UHFFFAOYSA-N
1-fmoc-piperidine-4-thiocarboxamide
98000101
9H-fluoren-9-ylmethyl 4-carbamothioylpiperidine-1-carboxylate
C1CN(CCC1C(=S)N)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
This item is not returnable. View return policy
941717-12-0
C21H22N2O2S
366.479
MFCD22988962
URQBPQRQAPSGOQ-UHFFFAOYSA-N
1-fmoc-piperidine-4-thiocarboxamide
98000101
9H-fluoren-9-ylmethyl 4-carbamothioylpiperidine-1-carboxylate
C1CN(CCC1C(=S)N)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 366.48
Percent Purity 97%
Chemical Name or Material 1-Fmoc-piperidine-4-thiocarboxamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'