missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2,3-dimethylimidazolium trifluoromethanesulfonate, 98%, Thermo Scientific™

Catalog Number p-7053362
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
174899-72-0
C8H13F3N2O3S
274.258
MFCD07784438
HMAHVRVKBSHMJX-UHFFFAOYSA-M
1-ethyl-2,3-dimethylimidazolium trifluoromethanesulfonate, 1-ethyl-2,3-dimethylimidazolium triflate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium trifluoromethanesulfonate, 3-ethyl-1,2-dimethylimidazol-1-ium triflate, dsstox_cid_29089, dsstox_rid_83308, dsstox_gsid_49233, 1-ethyl-2 3-dimethylimidazolium trifluor, 1-ethyl-2,3-dimethylimidazol-1-ium triflate
16210995
1-ethyl-2,3-dimethylimidazol-3-ium;trifluoromethanesulfonate
CCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[O-]
This item is not returnable. View return policy
174899-72-0
C8H13F3N2O3S
274.258
MFCD07784438
HMAHVRVKBSHMJX-UHFFFAOYSA-M
1-ethyl-2,3-dimethylimidazolium trifluoromethanesulfonate, 1-ethyl-2,3-dimethylimidazolium triflate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium trifluoromethanesulfonate, 3-ethyl-1,2-dimethylimidazol-1-ium triflate, dsstox_cid_29089, dsstox_rid_83308, dsstox_gsid_49233, 1-ethyl-2 3-dimethylimidazolium trifluor, 1-ethyl-2,3-dimethylimidazol-1-ium triflate
16210995
1-ethyl-2,3-dimethylimidazol-3-ium;trifluoromethanesulfonate
CCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 274.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Ethyl-2,3-dimethylimidazolium trifluoromethanesulfonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'