missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-difluorobenzene, 99%, Thermo Scientific™

Catalog Number p-3756988
Click to view available options
Quantity:
2.5 mL
10 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
461-96-1
C6H3BrF2
192.99
MFCD00010305
JHLKSIOJYMGSMB-UHFFFAOYSA-N
3,5-difluorobromobenzene, 1-bromo-3,5-difluoro-benzene, 1-bromo-3,5-difluoro benzene, benzene, 1-bromo-3,5-difluoro, 3,5-difluoro-bromobenzene, pubchem1042, acmc-1ae0k, 3,5-difluoro bromobenzene, bromo-3,5-difluorobenzene, 3,5-difluorophenyl bromide
136313
1-bromo-3,5-difluorobenzene
FC1=CC(Br)=CC(F)=C1
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
461-96-1
C6H3BrF2
192.99
MFCD00010305
JHLKSIOJYMGSMB-UHFFFAOYSA-N
3,5-difluorobromobenzene, 1-bromo-3,5-difluoro-benzene, 1-bromo-3,5-difluoro benzene, benzene, 1-bromo-3,5-difluoro, 3,5-difluoro-bromobenzene, pubchem1042, acmc-1ae0k, 3,5-difluoro bromobenzene, bromo-3,5-difluorobenzene, 3,5-difluorophenyl bromide
136313
1-bromo-3,5-difluorobenzene
FC1=CC(Br)=CC(F)=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -27.0°C
Color Colorless to Yellow
Density 1.6760g/mL
Boiling Point 140.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H3F2
Refractive Index 1.4980 to 1.5000
Quantity 2.5 mL
Beilstein 05, 679
Specific Gravity 1.676
Solubility Information Solubility in water: 0.238g/L (20°C)
Formula Weight 192.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromo-3, 5-difluorobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-3,5-difluorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'