missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-methylimidazolium hexafluorophosphate, 97%, Thermo Scientific™

Catalog Number p-7053288
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
433337-11-2
C11H13F6N2P
318.203
MFCD07784448
VVCQOUQQPZPIKL-UHFFFAOYSA-N
1-benzyl-3-methylimidazolium hexafluorophosphate, 1-benzyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 3-benzyl-1-methylimidazol-1-ium hexafluorophosphate, ksc943e0j, 1-benzyl-3-methylimid-azolium hexafluorophosphate, 1-methyl-3-benzylimidazolium hexafluorophosphate, 1-benzyl-3-methylimidazol-1-ium hexafluorophosphate, 1h-imidazolium, 1-methyl-3-phenylmethyl-, hexafluorophosphate 1-
11461285
1-benzyl-3-methylimidazol-3-ium;hexafluorophosphate
C[N+]1=CN(C=C1)CC2=CC=CC=C2.F[P-](F)(F)(F)(F)F
This item is not returnable. View return policy
433337-11-2
C11H13F6N2P
318.203
MFCD07784448
VVCQOUQQPZPIKL-UHFFFAOYSA-N
1-benzyl-3-methylimidazolium hexafluorophosphate, 1-benzyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 3-benzyl-1-methylimidazol-1-ium hexafluorophosphate, ksc943e0j, 1-benzyl-3-methylimid-azolium hexafluorophosphate, 1-methyl-3-benzylimidazolium hexafluorophosphate, 1-benzyl-3-methylimidazol-1-ium hexafluorophosphate, 1h-imidazolium, 1-methyl-3-phenylmethyl-, hexafluorophosphate 1-
11461285
1-benzyl-3-methylimidazol-3-ium;hexafluorophosphate
C[N+]1=CN(C=C1)CC2=CC=CC=C2.F[P-](F)(F)(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 318.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Benzyl-3-methylimidazolium hexafluorophosphate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'