Learn More
1-Adamantaneethanol, 98%
Product Code. 11307346
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11307346
Brand: Thermo Scientific Alfa Aesar L09318.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 6240-11-5 | C12H22O2 | 198.31 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6240-11-5
MDL Number MFCD00074756
Specifications
Melting Point 73°C to 75°C
Molecular Formula C12H22O2
Quantity 1 g
Beilstein 1855382
Synonym hexanoic acid, cyclohexyl ester, cyclohexyl caproate, unii-v3b77j8gnh, v3b77j8gnh, cyclohexyl n-hexanoate, hexanoic acid cyclohexyl ester, acmc-1b676, 4-4-methoxyphenyl-1,3-thiazol-4-amine
InChI Key DTNOERNOMHQUCN-UHFFFAOYSA-N
SMILES CCCCCC(=O)OC1CCCCC1
IUPAC Name cyclohexyl hexanoate
Molecular Weight (g/mol) 198.31
PubChem CID 80388
Formula Weight 180.29
Percent Purity 98%
Chemical Name or Material 1-Adamantaneethanol