missing translation for 'onlineSavingsMsg'
Learn More

1,9-Decadiyne, 97%, Thermo Scientific Chemicals

Catalog Number p-7021481
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1720-38-3
C10H14
134.22
MFCD00014926
ILVDYAGPHFWNQI-UHFFFAOYSA-N
1,9-decadiyne, acmc-1bppd, 1,9-decadiyne 6ci, 7ci, 8ci, 9ci
74400
CHEBI:37823
deca-1,9-diyne
C#CCCCCCCC#C
This item is not returnable. View return policy
1720-38-3
C10H14
134.22
MFCD00014926
ILVDYAGPHFWNQI-UHFFFAOYSA-N
1,9-decadiyne, acmc-1bppd, 1,9-decadiyne 6ci, 7ci, 8ci, 9ci
74400
CHEBI:37823
deca-1,9-diyne
C#CCCCCCCC#C

One-step synthesis of carbametacyclophane 3 from 1,9-decadiyne and diethyl acetylenedicarboxylate is described. The use of 1,9-decadiyne as a nucleophile led only to the monoalkynylation product, a selectivity that would be difficult to achieve with a catalytic in situ metalation process. 1,9-decadiyne reacts with iodobenzene (excess) to afford the bis-Sonogashira coupling product in excellent yield. 1,9-Decadiyne was dihydrated and gave 2,9-decanedione in 92% yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
One-step synthesis of carbametacyclophane 3 from 1,9-decadiyne and diethyl acetylenedicarboxylate is described. The use of 1,9-decadiyne as a nucleophile led only to the monoalkynylation product, a selectivity that would be difficult to achieve with a catalytic in situ metalation process. 1,9-decadiyne reacts with iodobenzene (excess) to afford the bis-Sonogashira coupling product in excellent yield. 1,9-Decadiyne was dihydrated and gave 2,9-decanedione in 92% yield.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.822
Boiling Point 80°C to 81°C (10 mmHg)
Flash Point 60°C (140°F)
Refractive Index 1.451
Quantity 5 g
UN Number UN3295
Beilstein 1734594
Solubility Information Insoluble in water.
Formula Weight 134.22
Percent Purity 97%
Chemical Name or Material 1,9-Decadiyne
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'