missing translation for 'onlineSavingsMsg'
Learn More

1,6-Naphthyridine-2-carboxylic acid, 98%, Thermo Scientific™

Catalog Number 15525256
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
197507-59-8
C9H6N2O2
174.16
OZZMWXQJCJUCEJ-UHFFFAOYSA-N
1,6-naphthyridine-2-carboxylicacid, 1,6 naphthyridine-2-carboxylic acid, peakdale1_000097, 2-1,6 naphthyridinecarboxylic acid, 2-carboxy-1,6-naphthyridine, pyridino 4,3-b pyridine-2-carboxylic acid
735908
1,6-naphthyridine-2-carboxylic acid
C1=CC(=NC2=C1C=NC=C2)C(=O)O
This item is not returnable. View return policy
197507-59-8
C9H6N2O2
174.16
OZZMWXQJCJUCEJ-UHFFFAOYSA-N
1,6-naphthyridine-2-carboxylicacid, 1,6 naphthyridine-2-carboxylic acid, peakdale1_000097, 2-1,6 naphthyridinecarboxylic acid, 2-carboxy-1,6-naphthyridine, pyridino 4,3-b pyridine-2-carboxylic acid
735908
1,6-naphthyridine-2-carboxylic acid
C1=CC(=NC2=C1C=NC=C2)C(=O)O
Melting Point 290.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 174.16
Percent Purity 98%
Chemical Name or Material 1,6-Naphthyridine-2-carboxylic acid
compliance-icons
Product Identifier
  • 1,6-Naphthyridine-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'