missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diaminoanthraquinone, tech. 90%, Thermo Scientific™

Product Code. p-7024460
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11464220

Brand: Thermo Scientific Alfa Aesar A19156.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Use as the intermediate of synthetic dyes. Use anthraquinone reductive dyes and dispersal dyes and acid dyes and intermediates. Itself to disperse dyestuffs purple. 1,4-Diaminoanthraquinone oxidized with different metal atoms gives rise to conducting coordination polymers consisting of difunctional ligand molecules linked by metal atoms. 1, 4-diaminoanthraquinone has solar cell applications. A hybrid Si/poly (1, 4-diaminoanthraquinone) photoconductive diode for optical sensor applications. Acts as the sequential incorporation of the mediator in electrochemical sensing of glucose on a gold electrode.

Solubility
Soluble in benzene, pyridine, the difficulties, slightly soluble in hot aniline acetate, ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-95-0
Molecular Formula C14H10N2O2
Molecular Weight (g/mol) 238.25
MDL Number MFCD00001224
InChI Key FBMQNRKSAWNXBT-UHFFFAOYSA-N
Synonym 1,4-diaminoanthraquinone, disperse violet 1, krisolamine, 9,10-anthracenedione, 1,4-diamino, grasol violet r, seacyl violet r, acetate red violet r, 1,4-diamino anthraquinone, setacyl violet r, oil violet r
PubChem CID 31420
IUPAC Name 1,4-diaminoanthracene-9,10-dione
SMILES NC1=CC=C(N)C2=C1C(=O)C1=CC=CC=C1C2=O

Specifications

Melting Point 260°C to 265°C
Odor Odorless
Quantity 25 g
Beilstein 2216556
Solubility Information Soluble in benzene,pyridine,the difficulties,slightly soluble in hot aniline acetate,ethanol.
Formula Weight 238.25
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,4-Diaminoanthraquinone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.