missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diiodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
1,3-Diiodopropane is used as a reagent for organic synthesis. 1, 3-Diiodopropane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Diiodopropane is used as a reagent for organic synthesis. 1, 3-Diiodopropane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Stable, but light sensitive - store in the dark. Incompatible with strong bases, strong oxidizing agents.
1,3-Diiodopropane is used as a reagent for organic synthesis. 1, 3-Diiodopropane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Stable, but light sensitive - store in the dark. Incompatible with strong bases, strong oxidizing agents.
Specifications
Melting Point | -20°C |
Density | 2.565 |
Boiling Point | 224°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.6415 |
Quantity | 50 g |
Beilstein | 1731117 |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 295.89 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'