missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluoroacetone, 98%, Thermo Scientific Chemicals

Catalog Number p-7033943
63.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
453-14-5
C3H4F2O
94.061
MFCD07368665
HKIPCXRNASWFRU-UHFFFAOYSA-N
1,3-difluoroacetone, 2-propanone, 1,3-difluoro, 1,3-difluoro-2-propanone, ch2f 2co, 1,3-bis fluoranyl propan-2-one, fluoromethyl ketone, sym-difluoroacetone, sgqdlibrbrwsp@, 1, 3-difluoroacetone, 1,3-difluoro acetone
228397
1,3-difluoropropan-2-one
C(C(=O)CF)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
453-14-5
C3H4F2O
94.061
MFCD07368665
HKIPCXRNASWFRU-UHFFFAOYSA-N
1,3-difluoroacetone, 2-propanone, 1,3-difluoro, 1,3-difluoro-2-propanone, ch2f 2co, 1,3-bis fluoranyl propan-2-one, fluoromethyl ketone, sym-difluoroacetone, sgqdlibrbrwsp@, 1, 3-difluoroacetone, 1,3-difluoro acetone
228397
1,3-difluoropropan-2-one
C(C(=O)CF)F

1,3-Difluoroacetone is used as pharmaceutical intermediate and for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Difluoroacetone is used as pharmaceutical intermediate and for chemical research.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Boiling Point 101°C to 102°C
Quantity 1 g
UN Number UN2929
Formula Weight 94.06
Percent Purity 98%
Chemical Name or Material 1,3-Difluoroacetone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'